Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit

Catalog No. BDB561521 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561521 100 Tests
BDB561523 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561521 Supplier BD Supplier No. 561521
Add to Cart
Add to Cart

Includes

Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Enhanced Sensitivity Detection Reagent (Part B), Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector

Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations

Detection reagents used in BD™ CBA Human Enhanced Sensitivity Flex Set System consist of two parts, Analyte Specific Detection Reagent (Part A), provided with the Flex Set and Enhanced Sensitivity Detection Reagent (Part B), provided in Master Buffer Kit with all other supporting reagents. Buffers, detection reagent and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.

  • Analyte working assay range: 274 to 200000fg/mL
  • Should not be used with Flex Sets from any other BD CBA System such as soluble protein, cell signalling etc
  • Available in pack sizes of 100 and 500 tests

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Includes Assay diluent, capture bead Diluent, detection reagent diluent, Wash Buffer, enhanced sensitivity detection reagent (Part B), instrument setup beads (A1, A9, F1, F9), PE instrument setup bead F1, PE positive control detector
Quantity 100 Tests
Content And Storage 2°C to 8°C
For Use With (Application) Flow Cytometry
Product Type Human enhanced sensitivity master Buffer kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.