Learn More
BD Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit
Shop All BD Biosciences ProductsDescription
Includes
Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Enhanced Sensitivity Detection Reagent (Part B), Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector
Detection reagents used in BD™ CBA Human Enhanced Sensitivity Flex Set System consist of two parts, Analyte Specific Detection Reagent (Part A), provided with the Flex Set and Enhanced Sensitivity Detection Reagent (Part B), provided in Master Buffer Kit with all other supporting reagents. Buffers, detection reagent and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.
- Analyte working assay range: 274 to 200000fg/mL
- Should not be used with Flex Sets from any other BD CBA System such as soluble protein, cell signalling etc
- Available in pack sizes of 100 and 500 tests
Flow Cytometry
Specifications
Specifications
| Cross Reactivity | Human |
| Includes | Assay diluent, capture bead Diluent, detection reagent diluent, Wash Buffer, enhanced sensitivity detection reagent (Part B), instrument setup beads (A1, A9, F1, F9), PE instrument setup bead F1, PE positive control detector |
| Quantity | 500 Tests |
| Content And Storage | 2°C to 8°C |
| Form | Buffer Solution |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human enhanced sensitivity master Buffer kit |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.