Learn More
Description
Includes
Standard, PE Detection Reagent, Capture Bead C7
- Capture bead and PE detection reagent formulated to 50x concentration
- Includes lyophilized standard
Specifications
Specifications
| Antigen | Eotaxin |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | 1 Vial capture bead 50X, 1 vial detection reagent 50X, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human eotaxin flex set (bead C7) |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.