Learn More
BD Cytometric Bead Array (CBA) Human Fractalkine; Flex Set, Bead C6
Shop All BD Biosciences ProductsDescription
Includes
Human Fractalkine, Standard; PE Detection Reagent; Capture Bead C6.
- Bead-based immunoassay
- Also known as CX3CL1
- Human reactivity determined by testing samples with BD CBA Human Fractalkine Flex
Specifications
Specifications
| Antigen | Fractalkine |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human Fractalkine, standard, PE detection reagent, capture bead C6 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human fractalkine flex set (bead C6) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.