Learn More
Description
Includes
- Human Granzyme A, Standard
- Human Granzyme A, PE Detection Reagent
- Human Granzyme A, Capture Bead D9
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. The Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 10,000 pg/mL.
- Human reactivity was determined by testing samples with BA Human Granzyme A Flex Set
- Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer, and FCAP Array Software
Specifications
Specifications
| Antigen | Granzyme B |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human Granzyme A, standard, Human Granzyme A, PE detection reagent, Human Granzyme A, capture bead D9 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human Granzyme A flex set (bead D9) |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.