Learn More
BD Cytometric Bead Array (CBA) Human IFN-γ Enhanced Sensitivity Flex Set (Bead B8)
Catalog No. BDB561515 Shop All BD Biosciences Products
Change view
Click to view available options
100 tests
Catalog No.
BDB561515 100 tests
1 options
This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Catalog No. BDB561515 Supplier BD Supplier No. 561515
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Includes
Kit contents:
  • Enhanced Sensitivity Capture Bead B8: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
  • Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
  • Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150
Measures human Human IFN-γ in serum, plasma and cell culture supernatant samples
This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.
  • Bead-based immunoassay
  • Reactivity: human
  • Standard assay range: 274 to 200000fg/mL
Flow cytometry/Immunoassay