Learn More
Description
Includes
Human IFNα; Standard; Human IFNα; PE Detection Reagent; Human IFNα; Capture Bead B8
The BD CBA Human IFNα Flex Set is a bead-based immunoassay capable of measuring soluble human IFN-α in serum, plasma, and cell culture supernatant samples
- Reactivity: QC Testing: Human
- Bead Position: B8
- Limit of detection is 1.5pg/mL
With BD CBA human soluble protein master buffer kit (Cat. No. 558264, 100 tests, or 558265, 500 tests), a flow cytometer, and the FCAP Array™ software
Specifications
Specifications
| Antigen | IFN-alpha |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human IFNa, standard, Human IFNa, PE detection reagent, Human IFNa, capture bead B8 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IFN-alpha flex set (bead B8) |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.