Learn More
Description
Includes
Human IFN-γ Standard; Human IFN-γ PE Detection Reagent; Human IFN-γ Capture Bead E7
Reactivity
- QC Testing: Human
- Testing in Development: Nonhuman Primate
Flow cytometry
Specifications
Specifications
| Antigen | IFN-γ |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human IFN-γ Standard, Human IFN-γ PE Detection Reagent, Human IFN-γ Capture Bead E7 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Powder |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IFN-γ Flex Set (Bead E7) |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.