Learn More
Description
Includes
Human IgE, Standard (two vials), Human total IgE PE Detection Reagent (one vial), Human total IgE Capture Bead E9 (one vial)
- Human reactivity was determined by testing samples with IgE Flex Set
- Theoretical limit of detection is 0.07ng/mL
- Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow cytometry
Specifications
Specifications
| Antigen | IgE |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human IgE, standard (2 vials), Human total IgE PE detection reagent (one vial), Human total IgE capture bead E9 (one vial) |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IgE flex set (bead E9) |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.