Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human IL-10; Flex Set, Bead B7

Catalog No. BDB558274 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558274 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558274 Supplier BD Supplier No. 558274
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Human IL-10; Standard; Human IL-10; PE Detection Reagent; Human IL-10; Capture Bead B7

Bead-based immunoassay capable of measuring human interleukin-10 (IL-10) in serum, plasma, and cell culture supernatant samples

Human reactivity was determined by testing samples with the BD CBA Human IL-10 Flex Set

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Conjugate PE
Cross Reactivity Human
Includes Human IL-10, standard, Human IL-10, PE detection reagent, Human IL-10, capture bead B7
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
Form Powder
For Use With (Application) Flow Cytometry
Product Type Human IL-10 flex set (bead B7)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.