Learn More
Description
Includes
Human IL-11; Standard; Human IL-11; PE Detection Reagent; Human IL-11; Capture Bead D5
Human reactivity was determined by testing samples with the BD CBA Human IL-11 Flex Set.
Specifications
Specifications
| Antigen | IL-11 |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Capture bead, detection reagent and standard |
| Quantity | 100 Tests |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-11 flex set (bead D5) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.