Learn More
BD Cytometric Bead Array (CBA) Human IL-12/IL-23p40; Flex Set, Bead E5
Shop All BD Biosciences ProductsDescription
Includes
Human IL-12; Standard; Human IL-12; PE Detection Reagent; Human IL-12; Capture Bead E5
- Capable of measuring the human p40 subunit of interleukin-12/Interluekin-23 (IL-12/IL-23p40) in serum, plasma, and cell culture supernatant samples
Specifications
Specifications
| Antigen | IL-12/IL-23p40 |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human IL-12, standard, Human IL-12, PE detection reagent, Human IL-12, capture bead E5 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-12/IL-23p40 flex set (bead E5) |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.