Learn More
Description
Includes
Human IL-12; Standard; Human IL-12; PE Detection Reagent; Human IL-12; Capture Bead E5
The BD CBA Human IL-12 p70 Flex Set is a bead based immunoassay capable of measuring human interleukin-12p70 (IL-12p70) in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human IL-12p70 Flex Set.
- Bead position: E5
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Specifications
Specifications
| Antigen | IL-12p70 |
| Conjugate | Unconjugated |
| Cross Reactivity | Human |
| Includes | Human IL-12, standard, Human IL-12, PE detection reagent, Human IL-12, capture bead E5 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-12p70 flex set (bead E5) |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.