Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human IL-17A Standard

Catalog No. BDB561668 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561668 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561668 Supplier BD Supplier No. 561668
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lyophilized recombinant Human Interleukin-17A (IL-17A) protein

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Approximates the CBA activity of Human IL-17A at a concentration of 2500 pg/mL when reconstituted in 4.0 mL assay diluent
  • Quantity: 100 tests

Flow cytometry/ Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Includes 1 Vial, lyophilized, Aqueous Buffered, reconstitute to 4mL
Quantity 100 Tests
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Flow Cytometry, Immunoassay
Product Type Human IL-17A standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.