Learn More
Description
Includes
Human IL-2; Standard; Human IL-2; PE Detection Reagent; Human IL-2; Capture Bead A4
Reactivity
- QC Testing: Human
- Testing in Development: Non-human Primate
Flow cytometry
Specifications
Specifications
| Antigen | IL-2 |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human IL-2, standard, Human IL-2, PE detection reagent, Human IL-2, capture bead A4 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-2 flex set (bead A4) |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.