Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human IL-21; Flex Set, Bead B8

Catalog No. BDB560358 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560358 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560358 Supplier BD Supplier No. 560358
Add to Cart
Add to Cart

Includes

Human IL-21; Standard; Human IL-21; PE Detection Reagent; Human IL-21; Capture Bead B8

Determines human reactivity by testing samples with BD CBA Human IL-21 Flex Set

The BD CBA Human IL-21 Flex Set is a bead-based immunoassay capable of measuring human Interleukin 21 (IL-21) in serum, plasma, and cell culture supernatant samples

  • Reactivity: QC Tested: Human
  • Bead Position: B8
  • Limit of detection: 34.3pg/mL

With BD CBA human soluble protein master buffer kit (Cat. No. 558264, 100 tests, or 558265, 500 tests), a flow cytometer, and the FCAP Array™ software

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-21
Cross Reactivity Human
Includes Human IL-21, standard, Human IL-21, PE detection reagent, Human IL-21, capture bead B8
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human IL-21 flex set (bead B8)

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.