Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human IL-3; Flex Set, Bead D5

Catalog No. BDB558355 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558355 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558355 Supplier BD Supplier No. 558355
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Human IL-3 Standard
  • Human IL-3, PE Detection Reagent
  • Human IL-3, Capture Bead D5

Bead-based immunoassay capable of measuring human interleukin-3 (IL-3) in serum, plasma, and cell culture supernatant samples

Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. Capture Bead and PE Detection Reagent components of this flex set have been formulated to 50x concentration to ensure product performance when multiplexed. Standard component is lyophilized and should be transferred to 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 2,500pg/mL.

  • Human reactivity was determined by testing samples with CBA Human IL-3 Flex Set
  • Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer and CAP Array Software

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-3
Cross Reactivity Human
Includes Human IL-3 standard, Human IL-3, PE detection reagent, Human IL-3, capture bead D5
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
Form Liquid
For Use With (Application) Flow Cytometry
Product Type Human IL-3 flex set (bead D5)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.