Learn More
BD Cytometric Bead Array (CBA) Human IL-5 Enhanced Sensitivity Flex Set (Bead A6)
Shop All Waters Corporation ProductsDescription
Includes
- Kit contents:
- Enhanced Sensitivity Capture Bead A6: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
- Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
- Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150
This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.
- Bead-based immunoassay
- Reactivity: human
- Standard assay range: 274 to 200000fg/mL
Flow cytometry/Immunoassay
Specifications
Specifications
| Content And Storage | Lyophilized in an aqueous buffered solution containing BSA™ and ProClin™ 150. Store at 4°C. |
| Includes | Capture beads, detection reagent, standards |
| For Use With (Application) | Used in conjunction with BD™ cytometric bead array (CBA) |
| No. of Tests | 100 |
| Quantity | 100 Tests |
| For Use With (Equipment) | Flow Cytometer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.