Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human IL-6 Enhanced Sensitivity Flex Set (Bead A7)

Catalog No. BDB561512 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561512 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Catalog No. BDB561512 Supplier BD Supplier No. 561512
Add to Cart
Add to Cart
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Includes

  • Kit contents:
  • Enhanced Sensitivity Capture Bead A7: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
  • Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
  • Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150

Measures human Interleukin-6 (IL-6) in serum, plasma and cell culture supernatant samples

This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.

  • Bead-based immunoassay
  • Reactivity: human
  • Standard assay range: 274 to 200000fg/mL

Applications:

Flow Cytometry/Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Cross Reactivity Human
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Human interleukin-6, flow cytometry and immunoassay
Product Type Human IL-6 enhanced sensitivity flex set (bead A7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.