Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human IL-8; Flex Set, Bead A9

Catalog No. BDB558277 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558277 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558277 Supplier BD Supplier No. 558277
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Human IL-8; Standard; Human IL-8; PE Detection Reagent; Human IL-8; Capture Bead A9

Bead-based immunoassay capable of measuring human interleukin-8 (IL-8), also know as CXCL8, in serum, plasma, and cell culture supernatant sample

Reactivity

  • QC Testing: Human
  • Testing in Development: Non-human Primate

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-8
Cross Reactivity Human
Includes Human IL-8, standard, Human IL-8, PE detection reagent, Human IL-8, capture bead A9
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human IL-8 flex set (bead A9)

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.