Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human Inflammatory Cytokine Kit Standard controls

Catalog No. bdb561666 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mL
1 Vial
Product Type:
Human Inflammatory Cytokine Standards
Human Th1/Th2/Th17 Cytokine Standards
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
BDB561666 1 Vial Human Th1/Th2/Th17 Cytokine Standards
BDB552932 0.2 mL Human Inflammatory Cytokine Standards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561666 Supplier BD Supplier No. 561666
Only null left
Add to Cart
Add to Cart

Have been tested to assure function as standards in CBA assays

  • Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
  • Should be reconstituted and serially diluted before use

Human Inflammatory Cytokine Standards

  • Contain mixture of recombinant human Interleukin (IL)-8, IL-1β, IL-6, IL-10, Tumor Necrosis Factor (TNF), and IL-12p70
  • Use with BD™ Cytometric Bead Array (CBA) Human Inflammatory Cytokine Kit
  • Size: 0.2mL
Human Th1/Th2/Th17 Cytokine Standards
  • Contain mixture of recombinant human Interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, Interferon-γ (IFN-γ), and Tumor Necrosis Factor (TNF)
  • Use with BD™ Cytometric Bead Array (CBA) Human Th1/Th2/TH17 Cytokine Kit
  • Size: 1 vial

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Includes Mixture of recombinant Human interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, interferon-g (IFN-g) and Tumor Necrosis Factor (TNF)
Quantity 1 Vial
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Flow Cytometry
Product Type Human Th1/Th2/Th17 Cytokine Standards
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.