Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human IP-10; Flex Set, Bead B5

Catalog No. BDB558280 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558280 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558280 Supplier BD Supplier No. 558280
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Human IP-10; Standard; Human IP-10; PE Detection Reagent; Human IP-10; Capture Bead B5

Bead-based immunoassay capable of measuring human interferon-Glc gamma-induced protein-10 (IP-10), also known as CXCL10, in serum, plasma, and cell culture supernatant samples

Reactivity

  • QC Testing: Human

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IP-10
Conjugate PE
Cross Reactivity Human
Includes Human IP-10, standard, Human IP-10, PE detection reagent, Human IP-10, capture bead B5
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
Form Powder
For Use With (Application) Flow Cytometry
Product Type Human IP-10 flex set (bead B5)

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.