Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human LTα; Flex Set, Bead D5

Catalog No. BDB560083 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560083 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560083 Supplier BD Supplier No. 560083
Add to Cart
Add to Cart

Includes

Human LTα, Standard; PE Detection Reagent; Capture Bead D5.

For measuring human lymphotoxin-α (LT-α) in serum, plasma, and cell culture supernatant samples

  • Bead-based immunoassay
  • Formerly known as tumor necrosis factor-β
  • Human reactivity determined by testing samples with BD CBA Human LT-α Flex

TRUSTED_SUSTAINABILITY

Specifications

Antigen LT-alpha
Cross Reactivity Human
Includes Human LTa, standard, PE detection reagent, capture bead D5
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human LT-alpha flex set (bead D5)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.