Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human MCP-1; Flex Set, Bead D8

Catalog No. BDB558287 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558287 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558287 Supplier BD Supplier No. 558287
Add to Cart
Add to Cart

Includes

Human MCP-1; Standard; Human MCP-1; PE Detection Reagent; Human MCP-1; Capture Bead D8

For detecting human monocyte chemoattractant proteins

The BD CBA Human MCP-1 Flex Set is a bead-based immunoassay capable of measuring human monocyte chemoattractant protein-1 (MCP-1), also known as CCL2, in serum, plasma, and cell culture supernatant samples. Human and non-human primate reactivity was determined by testing samples with the BD CBA Human MCP-1 Flex Set

  • Bead position: D8
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-1
Cross Reactivity Human
Includes Human MCP-1, standard, Human MCP-1, PE detection reagent, Human MCP-1, capture bead D8
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human MCP-1 flex set (bead D8)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.