Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human MIG; Flex Set, Bead E8

Catalog No. BDB558286 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558286 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558286 Supplier BD Supplier No. 558286
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Human MIG; Standard; Human MIG; PE Detection Reagent; Human MIG; Capture Bead E8

Bead-based immunoassay capable of measuring human monokine-induced by interferon-γ (MIG), also known as CXCL9, in serum, plasma, and cell culture supernatant samples

Human reactivity was determined by testing samples with the BD CBA Human MIG Flex Set.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MIG
Conjugate PE
Cross Reactivity Human
Includes Capture bead, detection reagent and standard
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Form Powder
For Use With (Application) Flow Cytometry
Product Type Human MIG flex set (bead E8)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.