Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human RANTES; Flex Set, Bead D4

Catalog No. BDB558324 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558324 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558324 Supplier BD Supplier No. 558324
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Human RANTES; Standard; Human RANTES; PE Detection Reagent; Human RANTES; Capture Bead D4

For detecting human RANTES, in serum, plasma, and cell culture samples

The BD CBA Human RANTES Flex Set is a bead-based immunoassay capable of measuring human RANTES, also known as CCL5, in serum, plasma, and cell culture supernatant samples. Human and non-human primate reactivity was determined by testing samples with the BD CBA Human RANTES Flex Set.

  • Reactivity: Human, Non-Human Primate
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen RANTES
Conjugate PE
Cross Reactivity Human
Includes Human RANTES, standard, Human RANTES, PE detection reagent, Human RANTES, capture bead D4
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Flow Cytometry
Product Type Human RANTES flex set (bead D4)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.