Learn More
Description
Includes
Human TGF-β1; Standard; Human TGF-β1; PE Detection Reagent; Human TGF-β1; Capture Bead B6
- Capable of measuring soluble human TGF-β1 in serum and cell culture supernatant samples
Specifications
Specifications
| Antigen | TGF-beta1 |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human TGF-beta1, standard, Human TGF-beta1, PE detection reagent, Human TGF-beta1, capture bead B6 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human TGF-beta1 single plex flex set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.