Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human TNF; Flex Set, Bead C4

Catalog No. BDB560112 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560112 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560112 Supplier BD Supplier No. 560112
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Human TNF; Standard; Human TNF; PE Detection Reagent; Human TNF; Capture Bead C4

Human and non-human primate reactivity was determined by testing samples with the BD CBA Human TNF Flex Set

  • Capable of measuring human tumor necrosis factor (TNF), formerly known as tumor necrosis factor-α, in serum, plasma, and cell culture supernatant samples

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Conjugate PE
Cross Reactivity Human
Includes Human TNF, standard, Human TNF, PE detection reagent, Human TNF, capture bead C4
Quantity 100 Tests
Content And Storage 4°C
Form Liquid
For Use With (Application) Flow Cytometry
Product Type Human TNF flex set (bead C4)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.