Learn More
Description
Includes
Human TNF; Standard; Human TNF; PE Detection Reagent; Human TNF; Capture Bead D9
Human and non-human primate reactivity was determined by testing samples with the BD CBA Human TNF Flex Set
Reactivity
- QC Testing: Human
- Testing in Development: Non-human Primate
Flow Cytometry
Specifications
Specifications
| Antigen | TNF |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Human TNF, standard, Human TNF, PE detection reagent, Human TNF, capture bead D9 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Powder |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human TNF flex set (bead D9) |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.