Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human TNFRI ; Flex Set, Bead C4

Catalog No. BDB560156 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560156 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560156 Supplier BD Supplier No. 560156
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Human TNFRI Standard; Human TNFRI PE Detection Reagent; Human TNFRI Capture Bead C4

Bead-based immunoassay for measuring soluble human type 1 tumor necrosis factor receptor (TNFRI), in serum, plasma and cell culture supernatant sample

Human reactivity was determined by testing samples with the BD™ CBA Human TNFRI Flex Set.

  • Reactivity: QC Tested: Human
  • Size: 100 tests

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFRI
Conjugate PE
Cross Reactivity Human
Includes Human TNFRI standard, Human TNFRI PE detection reagent, Human TNFRI capture bead C4
Quantity 100 Tests
Content And Storage 4°C
Form Liquid
For Use With (Application) Flow Cytometry
Product Type Human soluble TNFRI flex set (bead C4)

For Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.