Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human TNFRII; Flex Set, Bead C5

Catalog No. BDB560155 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560155 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560155 Supplier BD Supplier No. 560155
Add to Cart
Add to Cart

Includes

Human TNFRII; Standard; Human TNFRII; PE Detection Reagent; Human TNFRII; Capture Bead C5

Human reactivity was determined by testing samples with the BD CBA Human TNFRII Flex Set

  • Capable of measuring soluble human type 2 tumor necrosis factor receptor (TNFRII), also known as CD120b, in serum, plasma, and cell culture supernatant samples

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFRII
Cross Reactivity Human
Includes Human TNFRII, standard, Human TNFRII, PE detection reagent, Human TNFRII, capture bead C5
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human soluble TNFRII flex set (bead C5)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.