Learn More
BD Cytometric Bead Array (CBA) Mouse CD62E (E-Selectin); Flex Set, Bead E6
Shop All BD Biosciences ProductsDescription
Includes
- Mouse soluble CD62E (E-Selectin) Standard
- Mouse CD62E (E-Selectin) PE Detection Reagent
- Mouse CD62E (E-Selectin) Capture Bead E6
Cotains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. Standard component is lyophilized and should be transferred to 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL assay Diluent, the standard has a protein concentration of 10,000pg/mL.
- Mouse reactivity was determined by testing samples with this Flex set
- Must be used in conjunction with CBA Mouse/Rat Soluble Protein Master Buffer Kit, flow cytometer and FCAP Array Software
Specifications
Specifications
| Antigen | CD62E (E-Selectin) |
| Cross Reactivity | Mouse |
| Includes | Mouse soluble CD62E (E-Selectin) standard, Mouse CD62E (E-Selectin) PE detection reagent, Mouse CD62E (E-Selectin) capture bead E6 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Buffer Solution |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse CD62E (E-Selectin) flex set (bead E6) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.