Learn More
BD Cytometric Bead Array (CBA) Mouse Enhanced Sensitivity Master Buffer Kit
Shop All Waters Corporation ProductsDescription
Includes
Assay Diluent (150mL), Capture Bead Diluent (30mL), Detection Reagent Diluent A (15mL), Detection Reagent Diluent B (35mL), Wash Buffer (650mL)
Host Species:
Flow Cytometry
Specifications
Specifications
| Cross Reactivity | Mouse |
| Includes | Assay diluent 150mL, capture bead Diluent 30mL, detection reagent diluent A 15mL, detection reagent diluent B 35mL, Wash Buffer 650mL |
| Quantity | 100 Tests |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse enhanced sensitivity master Buffer kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.