Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse Enhanced Sensitivity Master Buffer Kit

Catalog No. BDB562248 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562248 500 Tests
BDB562246 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB562248 Supplier BD Supplier No. 562248
Add to Cart
Add to Cart

Includes

Assay Diluent (150mL), Capture Bead Diluent (30mL), Detection Reagent Diluent A (15mL), Detection Reagent Diluent B (35mL), Wash Buffer (650mL)

For use in Flow Cytometry

Host Species:

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Mouse
Includes Assay diluent 150mL, capture bead Diluent 30mL, detection reagent Diluent A 15mL, detection reagent diluent B 35mL, Wash Buffer 650mL
Quantity 500 Tests
Content And Storage 2°C to 8°C
For Use With (Application) Flow Cytometry
Product Type Mouse enhanced sensitivity master Buffer kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.