Learn More
Description
Includes
Mouse G-CSF; Standard; Mouse G-CSF; PE Detection Reagent; Mouse G-CSF; Capture Bead D4
- Capable of measuring mouse Granulocyte-Colony Stimulating Factor (G-CSF) in serum and cell culture supernatant samples
Specifications
Specifications
| Antigen | G-CSF |
| Cross Reactivity | Mouse |
| Includes | Mouse G-CSF, standard, Mouse G-CSF, PE detection reagent, Mouse G-CSF, capture bead D4 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse G-CSF flex set (bead D4) |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.