Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse GM-CSF; Flex Set, Bead B9

Catalog No. BDB558347 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558347 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558347 Supplier BD Supplier No. 558347
Add to Cart
Add to Cart

Includes

Mouse GM-CSF; Standard; Mouse GM-CSF; PE Detection Reagent; Mouse GM-CSF; Capture Bead B9

A bead-based immunoassay for measuring mouse granulocyte-macrophage colony stimulating factor (GM-CSF) in serum and cell culture supernatant samples

Mouse reactivity is determined by testing samples with the BD CBA Mouse GM-CSF Flex Set.

  • Reactivity: Mouse
  • Storage Buffer: Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen GM-CSF
Cross Reactivity Mouse
Includes Mouse GM-CSF, standard, Mouse GM-CSF, PE detection reagent, Mouse GM-CSF, capture bead B9
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse GM-CSF flex set (bead B9)

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.