Learn More
Description
Includes
Mouse IFN_γ; Standard; Mouse IFN_γ; PE Detection Reagent; Mouse IFN_γ; Capture Bead A4
The BD CBA Mouse IFN_γ Flex Set is a bead-based immunoassay capable of measuring mouse interferon_γ (IFN_γ) in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse IFN_γ Flex Set.
- Bead position: A4
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Specifications
Specifications
| Antigen | IFN-gamma |
| Cross Reactivity | Mouse |
| Includes | Mouse IFN-g, standard, Mouse IFNg, PE detection reagent, Mouse IFNg, capture bead A4 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Buffer Solution |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse IFN-gamma flex set (bead A4) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.