Learn More
Description
Includes
Mouse IL-1α; Standard; Mouse IL-1α; PE Detection Reagent; Mouse IL-1α; Capture Bead E4
The BD CBA Mouse IL-1α Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-1α (IL-1α) in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse IL-1α Flex Set.
- Reactivity: QC Tested: Mouse
- Bead Position: E4
- Limit of Detection: 0.8pg/mL
Specifications
Specifications
| Antigen | IL-1alpha |
| Conjugate | PE |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-1a, standard, Mouse IL-1a, PE detection reagent, Mouse IL-1a, capture bead E4 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse IL-1alpha flex set (bead E4) |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.