Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-1β; Flex Set, Bead E5

Catalog No. BDB560232 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560232 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560232 Supplier BD Supplier No. 560232
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Mouse IL-1β; Standard; Mouse IL-1β; PE Detection Reagent; Mouse IL-1β; Capture Bead E5

Determines mouse reactivity by testing samples with BD CBA Mouse IL-1β Flex Set

The BD CBA Mouse IL-1β Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-1β (IL-1β) in serum and cell culture supernatant samples

  • Reactivity: QC Tested: Mouse
  • Bead Position: E5
  • Limit of detection: 1.9pg/mL

With a BD CBA Mouse/Rat Soluble Protein Master Buffer Kit (Cat. No. 558266, 100 tests, or 558267, 500 tests), a flow cytometer, and the FCAP Array™ software

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1beta
Conjugate PE
Cross Reactivity Mouse
Includes Mouse IL-1beta, standard, Mouse IL-1beta, PE detection reagent, Mouse IL-1beta, capture bead E5
Quantity 100 Tests
Content And Storage 4°C
Form Liquid
For Use With (Application) Flow Cytometry
Product Type Mouse IL-1beta flex set (bead E5)

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.