Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-1β Standard

Catalog No. BDB561667 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561667 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561667 Supplier BD Supplier No. 561667

Contains lyophilized recombinant Mouse Interleukin-1β (IL-1β) protein

  • Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
  • For use with BD™ CBA Mouse IL-1β Flex Set
  • Approximates CBA activity of Mouse IL-1β at a concentration of 2500pg/mL when reconstituted in 4.0mL assay diluent
  • Size: 1 vial

Flow Cytometry/Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Mouse
Quantity 1 Vial
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Flow Cytometry, Immunoassay
Product Type Mouse IL-1beta standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.