Learn More
BD Cytometric Bead Array (CBA) Mouse IL-12/IL-23P40; Flex Set, Bead D7
Shop All BD Biosciences ProductsDescription
Includes
Mouse IL-12/IL-23P40, Standard (two vials), Mouse IL-12/IL-23P40 PE Detection Reagent (one vial), Mouse IL-12/IL-23P40; Capture Bead D7 (one vial)
- Mouse reactivity was determined by testing samples with Flex set
- Theoretical limit of detection is 13.6pg/mL
- Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow cytometry
Specifications
Specifications
| Antigen | IL-12/IL-23P40 |
| Conjugate | PE |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-12/IL-23P40, standard (2 vials), Mouse IL-12/IL-23P40 PE detection reagent (one vial), Mouse IL-12/IL-23P40, capture bead D7 (one vial) |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Buffer Solution |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse IL-12/IL-23p40 flex set (bead D7) |
Safety and Handling
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.