Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-17A; Enhanced Sensitivity Flex Set ,Bead C5

Catalog No. BDB562261 Shop All BD Biosciences Products
Change view
Click to view available options
100 tests;
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BDB562261 100 tests;
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB562261 Supplier BD Supplier No. 562261
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Mouse IL-17 Standard, Mouse IL-17 Enhanced Sensitivity Capture Bead C5, Mouse IL-17 Detection Reagent (Part A)

Sufficient for 100 tests

Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.