Learn More
Description
Includes
- Kit contents:
- Capture Bead D6: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
- PE Detection Reagent: stored in aqueous buffered solution containing BSA and ≤0.09% sodium azide
- Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Mouse/Rat Soluble Protein Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.
- Bead-based immunoassay
- Reactivity: mouse
- Top standard point: 2500pg/mL
Flow Cytometry/Immunoassay
Specifications
Specifications
| Antigen | IL-17F |
| Cross Reactivity | Mouse |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Measures Mouse interleukin-17F, flow cytometry and immunoassay |
| Product Type | Mouse IL-17F flex set (bead D6) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.