Learn More
Description
Includes
Mouse IL-21; Standard; Mouse IL-21; PE Detection Reagent; Mouse IL-21; Capture Bead B6
The BD™ CBA Mouse IL-21 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-21 (IL-21) in serum and cell culture supernatant samples
- Reactivity: QC Tested: Mouse
- Bead Position: B6
- Limit of detection is 4.8pg/mL
With BD CBA Mouse/Rat soluble protein master buffer kit (Cat. No. 558266, 100 tests, or 558267, 500 tests), flow cytometer, and FCAP Array™ software
Specifications
Specifications
| Antigen | IL-21 |
| Conjugate | PE |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-21, standard, Mouse IL-21, PE detection reagent, Mouse IL-21, capture bead B6 |
| Quantity | 100 Tests |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse IL-21 flex set (bead B6) |
Safety and Handling
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.