Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-23 p19/p40 Flex Set (Bead D7)

Catalog No. BDB562575 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562575 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Catalog No. BDB562575 Supplier BD Supplier No. 562575
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Includes

  • Kit contents:
  • Capture Bead D7: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
  • PE Detection Reagent: stored in aqueous buffered solution containing BSA and ≤0.09% sodium azide
  • Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150

Measures mouse Interleukin-23 p19/p40 (IL-23 p19/p40) in serum and cell culture supernatant samples

This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Mouse/Rat Soluble Protein Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.

  • Bead-based immunoassay
  • Reactivity: mouse
  • Top standard point: 2500pg/mL

Flow Cytometry/Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-23
Conjugate PE
Cross Reactivity Mouse
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Cell Culture Supernatant
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Measures Mouse interleukin-23 (p19/p40), flow cytometry
Product Type Mouse IL-23 p19/p40 flex set (bead D7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.