Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-4 Enhanced Sensitivity Flex Set (Bead A7)

Catalog No. BDB562272 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562272 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562272 Supplier BD Supplier No. 562272
Add to Cart
Add to Cart

Measures mouse Interleukin-4 (IL-4) in cell culture supernatant samples

  • Capable of measuring soluble analytes present in very low concentrations
  • Protein concentration of 200,000 fg/ml (when reconstituted in 0.4 ml Assay Diluent)

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4
Cross Reactivity Mouse
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Mouse interleukin-4, flow cytometry
Product Type Mouse IL-4 enhanced sensitivity flex set (bead A7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.