Learn More
Description
Includes
Mouse IL-6; Standard; Mouse IL-6; PE Detection Reagent; Mouse IL-6; Capture Bead B4
The BD CBA Mouse IL-6 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-6 (IL-6) in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse IL-6 Flex Set.
- Reactivity: Mouse
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Specifications
Specifications
| Antigen | IL-6 |
| Conjugate | PE |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-6, standard, Mouse IL-6, PE detection reagent, Mouse IL-6, capture bead B4 |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Buffer Solution |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse IL-6 flex set (bead B4) |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.