Learn More
Description
Includes
Mouse IL-9 Standard (two vials), Mouse IL-9 PE Detection Reagent (one vial), Mouse IL-9 Capture Bead B5 (one vial)
- Mouse reactivity was determined by testing samples with Flex Set
- Theoretical limit of detection is 10.7 units/mL
- Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow cytometry
Specifications
Specifications
| Antigen | IL-9 |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-9 standard (2 vials), Mouse IL-9 PE detection reagent (one vial), Mouse IL-9 capture bead B5 (one vial) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse IL-9 flex set (bead B5) |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.