Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse MCP-1; Flex Set, Bead B7

Catalog No. BDB558342 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558342 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558342 Supplier BD Supplier No. 558342
Add to Cart
Add to Cart

Includes

Mouse MCP-1; Standard; Mouse MCP-1; PE Detection Reagent; Mouse MCP-1; Capture Bead B7

For detecting mouse monocyte chemoattractant protein-1 in serum and cell culture samples

The BD CBA Mouse MCP-1 Flex Set is a bead-based immunoassay capable of measuring mouse monocyte chemoattractant protein-1 (MCP-1), also known as CCL2, in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse MCP-1 Flex Set.

  • Reactivity: Mouse
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-1
Cross Reactivity Mouse
Includes Mouse MCP-1, standard, Mouse MCP-1, PE detection reagent, Mouse MCP-1, capture bead B7
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse MCP-1 flex set (bead B7)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.