Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse MIP-1α; Flex Set, Bead C7

Catalog No. BDB558449 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558449 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558449 Supplier BD Supplier No. 558449
Add to Cart
Add to Cart

Includes

Mouse MIP-1α, Standard (two vials), Mouse MIP-1α PE Detection Reagent (one vial), Mouse MIP-1α Capture Bead C7 (one vial)

Bead-based immunoassay for measuring mouse Macrophage Inflammatory Protein-1α (MIP-1α) in serum and cell culture supernatant samples

  • Mouse reactivity was determined by testing samples with Flex Set
  • Theoretical limit of detection is 2.3 pg/mL
  • Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-1alpha
Cross Reactivity Mouse
Includes Mouse MIP-1a, standard (2 vials), Mouse MIP-1a PE detection reagent (one vial), Mouse MIP-1a capture bead C7 (one vial)
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse MIP-1alpha flex set (bead C7)

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.